3-Fluoro-4-formylpyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Fluoro-4-pyridinecarbaldehyde, 3-Fluoroisonicotinaldehyde, 3-Fluoro-4-pyridinecarboxaldehyde
064643-1G
064643-5G
101928-094EA
116.92
USD
101928-094
101928-096
3-Fluoro-4-formylpyridine ≥97%
3-Fluoro-4-formylpyridine
Hazard code(s): Irritant, Refrigerate
Formula:
C₆H₄FNO MW: 125.1 g/mol |
MDL Number:
MFCD06410686 CAS Number: 40273-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...