5-Nitro-o-toluic acid
Supplier: Matrix Scientific
064570-5G
064570-25G
064570-100G
101927-944EA
23.08
USD
101927-944
101927-946
102551-650
5-Nitro-o-toluic acid
5-Nitro-o-toluic acid
Formula:
C₈H₇NO₄ Melting Pt: 179…181 °C Density: 1.49 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007371 CAS Number: 1975-52-6 EINECS: 217-829-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...