Indole-3-carbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
1H-Indole-3-carbonitrile, 3-Cyano-1H-indole, 3-Cyanoindole
064203-1G
064203-5G
101927-258EA
15.38
USD
101927-258
101927-260
Indole-3-carbonitrile ≥95%
Indole-3-carbonitrile
Hazard code(s): Irritant
Formula:
C₉H₆N₂ MW: 142.16 g/mol Boiling Pt: 350 °C (1013 hPa) |
MDL Number:
MFCD00022717 CAS Number: 5457-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...