1-Methyl-3-piperidinemethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Hydroxymethyl)-1-methylpiperidine

064163-25G 064163-100G
101927-176EA 38.46 USD
101927-176 101927-178
1-Methyl-3-piperidinemethanol ≥95%
1-Methyl-3-piperidinemethanol

Hazard code(s): Irritant

Formula: C₇H₁₅NO
MW: 129.2 g/mol
Boiling Pt: 140…145 °C
Density: 1.013 g/cm³
MDL Number: MFCD00006497
CAS Number: 7583-53-1
EINECS: 231-488-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR