1-(2-Nitrophenyl)piperazine

Supplier: Matrix Scientific

064036-1G 064036-5G
101926-864EA 23.08 USD
101926-864 101926-866
1-(2-Nitrophenyl)piperazine
1-(2-Nitrophenyl)piperazine

Hazard code(s): Irritant-Harmful

Formula: C₁₀H₁₃N₃O₂
MW: 207.23 g/mol
Boiling Pt: 142…144 °C (1mm)
MDL Number: MFCD00040728
CAS Number: 59084-06-9
EINECS: 261-593-1

Order Now

Specification Test Results

Boiling point 142-144 °C/

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR