2,4,6-Trimethyl-m-phenylenediamine for synthesis

Supplier: Matrix Scientific

Synonyms: 1,3-Diamino-2,4,6-trimethylbenzene, 2,4-Diamino-1,3,5-trimethylbenzene, 2,4,6-Trimethyl-1,3-phenylenediamine, 2,4-Diaminomesitylene, 2,4,6-Trimethylbenzene-1,3-diamine

063980-5G 063980-25G
101926-756EA 16.92 USD
101926-756 101926-758
2,4,6-Trimethyl-m-phenylenediamine for synthesis
2,4,6-Trimethyl-m-phenylenediamine
Formula: C₉H₁₄N₂
MDL Number: MFCD00010149
CAS Number: 3102-70-3
EINECS: 221-456-9
UN: 2811
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR