3-Amino-2-naphthol

Supplier: Matrix Scientific

Synonyms: 3-Amino-2-hydroxynaphthalene, 3-Aminonaphthalen-2-ol

063963-1G 063963-5G 063963-25G
101926-718EA 72.31 USD
101926-718 101926-720 101926-722
3-Amino-2-naphthol
3-Amino-2-naphthol

Hazard code(s): Irritant

Formula: C₁₀H₉NO
MW: 159.19 g/mol
Melting Pt: 232…238 °C
MDL Number: MFCD00004113
CAS Number: 5417-63-0
EINECS: 226-519-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR