4,5-Dicyano-1-methylimidazole

Supplier: Matrix Scientific

Synonyms: 4,5-Dicyano-1-methyl-1H-imidazole, 1-Methyl-1H-imidazole-4,5-dicarbonitrile, 1-Methylimidazole-4,5-dicarbonitrile

062727-1G 062727-5G
101924-440EA 144.62 USD
101924-440 101924-442
4,5-Dicyano-1-methylimidazole
4,5-Dicyano-1-methylimidazole
Formula: C₆H₄N₄
MW: 132.12 g/mol
MDL Number: MFCD01567299
CAS Number: 19485-35-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR