2-Methylaminoethanol

Supplier: Matrix Scientific
Danger

Synonyms: N-Methyl-2-ethanolamin, N-Methyl-2-aminoethanol, 2-(Methylamino)ethanol

062563-100G 062563-500G
101924-116EA 38.46 USD
101924-116 101924-118
2-Methylaminoethanol
2-Methylaminoethanol

Hazard code(s): Corrosive

Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 159 °C
Melting Pt: –5 °C
Density: 0.937 g/cm³ (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00002839
CAS Number: 109-83-1
EINECS: 203-710-0
Merck Index: 12,06096

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR