3-Chloro-6-methylpyridazine

Supplier: Matrix Scientific

062540-5G 062540-25G 062540-100G
101924-068EA 92.31 USD
101924-068 101924-070 102551-586
3-Chloro-6-methylpyridazine
3-Chloro-6-methylpyridazine
Formula: C₅H₅ClN₂
MDL Number: MFCD00052905
CAS Number: 1121-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR