2-Amino-4-chloro-6-methylpyrimidine ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-6-methylpyrimidin-2-amine

059900-25G 059900-100G
101919-430EA 27.69 USD
101919-430 101919-432
2-Amino-4-chloro-6-methylpyrimidine ≥98%
2-Amino-4-chloro-6-methylpyrimidine

Hazard code(s): Irritant, AVOID SKIN CONTACT

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
Melting Pt: 181…186 °C
MDL Number: MFCD00006091
CAS Number: 5600-21-5
EINECS: 227-018-3

Order Now

Specification Test Results

Melting point 181-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR