2-Bromo-3-(trifluoromethyl)aniline ≥97% for synthesis

Supplier: Matrix Scientific

059669-5G 059669-25G
101918-848EA 153.85 USD
101918-848 102833-284
2-Bromo-3-(trifluoromethyl)aniline ≥97% for synthesis
2-Bromo-3-(trifluoromethyl)aniline

Hazard code(s): Irritant, Light Sensitive, Refrigerate

Formula: C₇H₅BrF₃N
MW: 240.02
MDL Number: MFCD01656033
CAS Number: 58458-10-9
EINECS: 261-261-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR