1-(2-Aminophenyl)piperazine ≥97%

Supplier: Matrix Scientific

059607-250MG 059607-1G
101918-632EA 121.54 USD
101918-632 101918-634
1-(2-Aminophenyl)piperazine ≥97%
1-(2-Aminophenyl)piperazine

Hazard code(s): Corrosive

Formula: C₁₀H₁₅N₃
Melting Pt: 106…116 °C
MDL Number: MFCD03444413
CAS Number: 13339-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR