(4-Chlorophenyl)acetic acid

Supplier: Matrix Scientific
Warning

059325-100G 059325-250G
101917-918EA 73.85 USD
101917-918 101917-920
(4-Chlorophenyl)acetic acid
(4-Chlorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₄Cl)CH₂COOH
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 103…105 °C
Density: 1.43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004344
CAS Number: 1878-66-6
EINECS: 217-521-6

Order Now

Specification Test Results

Melting point 103-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR