4-Iodo-3,5-xylenol ≥95%

Supplier: Matrix Scientific

059107-5G 059107-25G
101917-116EA 263.08 USD
101917-116 101917-118
4-Iodo-3,5-xylenol ≥95%
4-Iodo-3,5-xylenol

Hazard code(s): Irritant

Formula: C₈H₉IO
MDL Number: MFCD00174284
CAS Number: 80826-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR