4-Hydroxy-3H-furan-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4-hydroxy-2-furanone, 4-hydroxyfuranone, 4-Hydroxyfuran-2(3H)-one

059087-1G 059087-5G
101917-024EA 149.23 USD
101917-024 101917-026
4-Hydroxy-3H-furan-2-one ≥95%
4-Hydroxy-3H-furan-2-one

Hazard code(s): Irritant

Formula: C₄H₄O₃
MW: 100.073 g/mol
MDL Number: MFCD11227258
CAS Number: 436162-45-7
EINECS: 208-785-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR