(1-Ethoxyethylidene)malononitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1-Ethoxyethylidene)malononitrile, 2-(1-Ethoxy-ethylidene)-malononitrile

059017-25G 059017-100G
101916-714EA 90.77 USD
101916-714 101916-716
(1-Ethoxyethylidene)malononitrile ≥95%
(1-Ethoxyethylidene)malononitrile

Hazard code(s): Toxic

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 89…93 °C
MDL Number: MFCD00001852
CAS Number: 5417-82-3
EINECS: 226-521-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR