2,4-Difluorophenylhydrazine ≥95%

Supplier: Matrix Scientific

058978-5G 058978-25G
101916-566EA 115.38 USD
101916-566 101916-568
2,4-Difluorophenylhydrazine ≥95%
2,4-Difluorophenylhydrazine

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₆F₂N₂
Melting Pt: 65…67 °C
MDL Number: MFCD00042256
CAS Number: 40594-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR