3,6-Dichloro-4-methylpyridazine ≥95%

Supplier: Matrix Scientific
Warning

058971-1G 058971-5G 058971-25G
101916-534EA 32.31 USD
101916-534 101916-536 101916-538
3,6-Dichloro-4-methylpyridazine ≥95%
3,6-Dichloro-4-methylpyridazine

Hazard code(s): Irritant

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 130…135 °C (1 mmHg)
Melting Pt: 87 °C
MDL Number: MFCD00006465
CAS Number: 19064-64-3
EINECS: 242-794-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR