3-(Chloromethyl)benzoyl chloride ≥95%
Supplier: Matrix Scientific
058862-5G
058862-25G
101916-090EA
52.31
USD
101916-090
101916-092
3-(Chloromethyl)benzoyl chloride ≥95%
3-(Chloromethyl)benzoyl chloride
Hazard code(s): Corrosive
Formula:
C₈H₆Cl₂O Boiling Pt: 149…150 °C (20mm) Melting Pt: 4 °C Density: 1.330 g/cm³ |
MDL Number:
MFCD00000682 CAS Number: 63024-77-1 EINECS: 263-799-7 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...