5-Aminobenzotriazole ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 1H-Benzo[d][1,2,3]triazol-5-amine, 5-Amino-1H-benzotriazole

058705-1G 058705-5G 058705-10G
101915-494EA 116.92 USD
101915-494 101915-496 101915-498
5-Aminobenzotriazole ≥95% for synthesis
5-Aminobenzotriazole

Hazard code(s): Irritant

Formula: C₆H₆N₄
MDL Number: MFCD00047209
CAS Number: 3325-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR