3-Fluoro-o-toluonitrile ≥98%

Supplier: Matrix Scientific

058533-5G 058533-25G
101915-030EA 129.23 USD
101915-030 101915-032
3-Fluoro-o-toluonitrile ≥98%
3-Fluoro-o-toluonitrile

Hazard code(s): Toxic, Irritant

Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD03788541
CAS Number: 185147-06-2
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR