3-Chloro-2-nitrobenzoic acid ≥97% for synthesis

Supplier: Matrix Scientific
Warning

058520-25G 058520-100G
101915-026EA 104.62 USD
101915-026 101915-028
3-Chloro-2-nitrobenzoic acid ≥97% for synthesis
3-Chloro-2-nitrobenzoic acid

Hazard code(s): Irritant-Harmful

Formula: C₇H₄ClNO₄
Melting Pt: 236 - 240 °C
Density: 1.566 g/cm³ (18 °C)
MDL Number: MFCD00007087
CAS Number: 4771-47-5
EINECS: 225-313-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR