4-Amino-2-chlorobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

058517-25G 058517-100G
101915-018EA 41.54 USD
101915-018 101915-020
4-Amino-2-chlorobenzoic acid ≥98%
4-Amino-2-chlorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00007772
CAS Number: 2457-76-3
EINECS: 219-540-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR