4,4'-Dibromobiphenyl (PBB No. 15)

Supplier: Matrix Scientific
Warning

Synonyms: PBB No. 15

058382-100G 058382-500G
101914-698EA 176.92 USD
101914-698 101914-700
4,4'-Dibromobiphenyl (PBB No. 15)
4,4'-Dibromobiphenyl (PBB No. 15)

Hazard code(s): Possible Carcinogen, Irritant

Formula: C₁₂H₈Br₂
MW: 312 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 164…169 °C
Density: 1.867 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000101
CAS Number: 92-86-4
EINECS: 202-198-6
UN: 3152
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR