6-Methyluracil

Supplier: Matrix Scientific

058315-100G 058315-500G
101914-568EA 35.38 USD
101914-568 101914-570
6-Methyluracil
6-Methyluracil
Formula: C₅H₆N₂O₂
Boiling Pt: 358 °C (1013 hPa)
Melting Pt: 117 °C
MDL Number: MFCD00006028
CAS Number: 626-48-2
EINECS: 210-949-4
Merck Index: 12,06213

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR