Diethyl acetamidomalonate
Supplier: Matrix Scientific
058312-100G
058312-500G
101914-562EA
30.77
USD
101914-562
101914-564
Diethyl acetamidomalonate
Diethyl acetamidomalonate
Formula:
C₉H₁₅NO₅ |
MDL Number:
MFCD00009146 CAS Number: 1068-90-2 EINECS: 213-952-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...