3-Quinuclidinone hydrochloride

Supplier: Matrix Scientific

Synonyms: 1-Azabicyclo[2.2.2]octan-3-one hydrochloride

058305-5G 058305-25G 058305-100G
101914-550EA 29.23 USD
101914-550 101914-552 101914-554
3-Quinuclidinone hydrochloride
3-Quinuclidinone hydrochloride
Formula: C₇H₁₂ClNO
MDL Number: MFCD00137391
CAS Number: 1193-65-3
EINECS: 214-776-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR