Methyl-2-chlorobenzoate ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Chlorobenzoic acid emthyl ester
057135-25G
057135-500G
101912-494EA
69.23
USD
101912-494
101912-498
Methyl-2-chlorobenzoate ≥98%
Methyl-2-chlorobenzoate
Hazard code(s): Irritant
Formula:
C₈H₇ClO₂ MW: 170.6 g/mol Boiling Pt: 234 °C Density: 1.247 g/cm³ (20 °C) |
MDL Number:
MFCD00016337 CAS Number: 610-96-8 EINECS: 210-242-0 |
Specification Test Results
Boiling point | 234°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...