Ethyl-3-nitrobenzoate ≥99%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-nitrobenzoate, 3-Nitrobenzoic acid ethyl ester

057074-25G 057074-100G
101912-268EA 176.92 USD
101912-268 101912-270
Ethyl-3-nitrobenzoate ≥99%
Ethyl-3-nitrobenzoate

Hazard code(s): Irritant

Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 41…42 °C
MDL Number: MFCD00014702
CAS Number: 618-98-4
EINECS: 210-574-6

Order Now

Specification Test Results

Melting point 41-42°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR