3-Ethoxybenzonitrile ≥98%

Supplier: Matrix Scientific

057060-10G 057060-50G
101912-218EA 232.31 USD
101912-218 101912-220
3-Ethoxybenzonitrile ≥98%
3-Ethoxybenzonitrile

Hazard code(s): Irritant

Formula: C₉H₉NO
MW: 147.18
Boiling Pt: 241…244 °C
Density: 1.05 g/cm³
MDL Number: MFCD00001805
CAS Number: 25117-75-3
EINECS: 246-632-2
UN: 3276
ADR: 6.1 III

Order Now

Specification Test Results

Boiling point 241-244°C
Density 1.05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR