4'-Ethoxyacetophenone ≥99%

Supplier: Matrix Scientific

057057-25G 057057-100G
101912-206EA 90.77 USD
101912-206 101912-208
4'-Ethoxyacetophenone ≥99%
4'-Ethoxyacetophenone

Hazard code(s): Irritant

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 83 °C (0.04 mmHg)
Melting Pt: 37…39 °C
MDL Number: MFCD00009095
CAS Number: 1676-63-7
EINECS: 216-825-6

Order Now

Specification Test Results

Melting point 37-39°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR