1-(3,5-Dimethylphenyl)-2-thiourea
Supplier: Matrix Scientific
|
Danger
|
Synonyms:
3,5-Dimethylphenylthiourea, N-(3,5-Dimethylphenyl)thiourea, 1-(3,5-Dimethylphenyl)thiourea, 1-(3,5-Xylyl)thiourea
057048-2G
057048-10G
101912-174EA
84.62
USD
101912-174
101912-176
1-(3,5-Dimethylphenyl)-2-thiourea
1-(3,5-Dimethylphenyl)-2-thiourea
Hazard code(s): Irritant
Formula:
(C₆H₃(CH₃)₂)NC(S)NH₂ MW: 180.27 g/mol Boiling Pt: 310 °C (1013 hPa) Melting Pt: 172 - 173 °C |
MDL Number:
MFCD00041173 CAS Number: 97480-60-9 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 171-173°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...