1-(2,3-Dimethylphenyl)-2-thiourea

Supplier: Matrix Scientific

Synonyms: 1-(2,3-Xylyl)thiourea, N-(2,3-Dimethylphenyl)thiourea, 2,3-Dimethylphenylthiourea

057047-2G 057047-10G
101912-170EA 83.08 USD
101912-170 101912-172
1-(2,3-Dimethylphenyl)-2-thiourea
1-(2,3-Dimethylphenyl)-2-thiourea

Hazard code(s): Irritant

Formula: (C₆H₃(CH₃)₂)NC(S)NH₂
MW: 180.27 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 202…204 °C (dec.)
MDL Number: MFCD00041163
CAS Number: 55752-58-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 202-204°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR