(2,5-Dimethylphenyl)acetonitrile ≥98%
Supplier: Matrix Scientific
057040-10G
057040-50G
101912-138EA
238.46
USD
101912-138
101912-140
(2,5-Dimethylphenyl)acetonitrile ≥98%
(2,5-Dimethylphenyl)acetonitrile
Hazard code(s): Irritant
Formula:
C₁₀H₁₁N Boiling Pt: 73 °C (1mm) Melting Pt: 27…29 °C |
MDL Number:
MFCD00013816 CAS Number: 16213-85-7 EINECS: 240-339-3 UN: 3439 ADR: 6.1,III |
Specification Test Results
Boiling point | 73°C/1mm |
Melting point | 27-29°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...