4-Methylveratrole ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: 1,2-Dimethoxy-4-methylbenzene, 3,4-Dimethoxytoluene

057032-5G 057032-25G
101912-112EA 73.85 USD
101912-112 101912-114
4-Methylveratrole ≥99%
4-Methylveratrole

Hazard code(s): Irritant

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 122 °C (27mm)
Melting Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00016651
CAS Number: 494-99-5
EINECS: 207-796-0

Order Now

Specification Test Results

Boiling point 122°C/27mm
Melting point 24°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR