(3,4-Dimethoxyphenyl)acetic acid hydrazide

Supplier: Matrix Scientific

057030-5G 057030-25G
101912-104EA 83.08 USD
101912-104 101912-106
(3,4-Dimethoxyphenyl)acetic acid hydrazide
(3,4-Dimethoxyphenyl)acetic acid hydrazide

Hazard code(s): Irritant

Formula: C₁₀H₁₄N₂O₃
MW: 210.23 g/mol
Melting Pt: 105…107 °C
MDL Number: MFCD00051701
CAS Number: 60075-23-2

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR