4,5-Dimethoxy-2-nitrobenzyl alcohol

Supplier: Matrix Scientific

Synonyms: 6-Nitroveratryl alcohol, (4,5-Dimethoxy-2-nitrophenyl)methanol

057029-5G 057029-25G
101912-100EA 113.85 USD
101912-100 101912-102
4,5-Dimethoxy-2-nitrobenzyl alcohol
4,5-Dimethoxy-2-nitrobenzyl alcohol

Hazard code(s): Irritant

Formula: C₉H₁₁NO₅
MW: 213.19 g/mol
Melting Pt: 145…148 °C
MDL Number: MFCD00014701
CAS Number: 1016-58-6
EINECS: 213-806-4

Order Now

Specification Test Results

Melting point 145-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR