2,5-Dimethoxybenzonitrile ≥98%

Supplier: Matrix Scientific

057026-5G 057026-25G
101912-092EA 87.69 USD
101912-092 101912-094
2,5-Dimethoxybenzonitrile ≥98%
2,5-Dimethoxybenzonitrile

Hazard code(s): Irritant

Formula: C₉H₉NO₂
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 81…83 °C
MDL Number: MFCD00016375
CAS Number: 5312-97-0
EINECS: 226-169-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 81-83°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR