2',4'-Dimethoxyacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(2,4-Dimethoxyphenyl)ethan-1-one

057024-25G 057024-100G
101912-084EA 44.62 USD
101912-084 101912-086
2',4'-Dimethoxyacetophenone ≥98%
2',4'-Dimethoxyacetophenone

Hazard code(s): Irritant

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 288 °C
Melting Pt: 39…41 °C
MDL Number: MFCD00008727
CAS Number: 829-20-9
EINECS: 212-587-2

Order Now

Specification Test Results

Boiling point 288°C
Melting point 39-41°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR