1-(2,5-Difluorophenyl)-2-thiourea

Supplier: Matrix Scientific

Synonyms: N-(2,5-Difluorophenyl)thiourea, 2,5-Difluorophenylthiourea, 1-(2,5-Difluorophenyl)thiourea

057019-2G 057019-10G
101912-064EA 112.31 USD
101912-064 101912-066
1-(2,5-Difluorophenyl)-2-thiourea
1-(2,5-Difluorophenyl)-2-thiourea

Hazard code(s): Irritant

Formula: (C₆H₃F₂)NC(S)NH₂
MW: 188.2 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 137 - 138 °C
MDL Number: MFCD00041152
CAS Number: 207981-44-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 137-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR