1-(3,5-Dichlorophenyl)-2-thiourea
Supplier: Matrix Scientific
|
Danger
|
057014-1G
057014-5G
101912-048EA
95.38
USD
101912-048
101912-050
1-(3,5-Dichlorophenyl)-2-thiourea
1-(3,5-Dichlorophenyl)-2-thiourea
Hazard code(s): Irritant
Formula:
(C₆H₃Cl₂)NC(S)NH₂ Boiling Pt: 326 °C (1013 hPa) Melting Pt: 180 °C |
MDL Number:
MFCD00041168 CAS Number: 107707-33-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 175-177°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...