1-(2,5-Dichlorophenyl)-2-thiourea

Supplier: Matrix Scientific

Synonyms: 2,5-Dichlorophenylthiourea, 1-(2,5-Dichlorophenyl)thiourea, N-(2,5-Dichlorophenyl)thiourea

057012-5G 057012-25G
101912-040EA 149.23 USD
101912-040 101912-042
1-(2,5-Dichlorophenyl)-2-thiourea
1-(2,5-Dichlorophenyl)-2-thiourea

Hazard code(s): Irritant

Formula: (C₆H₃Cl₂)NC(S)NH₂
MW: 221.11 g/mol
Melting Pt: 320 °C
MDL Number: MFCD00041154
CAS Number: 4949-85-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 192-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR