2,5-Dichloro-3-nitrobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

057008-2G 057008-10G
101912-026EA 315.38 USD
101912-026 101912-028
2,5-Dichloro-3-nitrobenzoic acid ≥98%
2,5-Dichloro-3-nitrobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₃Cl₂NO₄
MW: 236.01 g/mol
Melting Pt: 224…226 °C
MDL Number: MFCD00014691
CAS Number: 88-86-8
EINECS: 201-862-2

Order Now

Specification Test Results

Melting point 224-226°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR