2,5-Dichlorobenzhydrazide ≥98%

Supplier: Matrix Scientific

Synonyms: 2,5-Dichlorobenzoic hydrazide

056999-5G 056999-25G
101912-010EA 130.77 USD
101912-010 101912-012
2,5-Dichlorobenzhydrazide ≥98%
2,5-Dichlorobenzhydrazide

Hazard code(s): Irritant

Formula: C₇H₆Cl₂N₂O
MW: 205.04 g/mol
Melting Pt: 180…181 °C
MDL Number: MFCD00014754
CAS Number: 67487-35-8

Order Now

Specification Test Results

Melting point 180-181°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR