2,4-Dichlorobenzhydrazide

Supplier: Matrix Scientific

Synonyms: 2,4-Dichlorobenzoic hydrazide

056998-10G 056998-50G
101912-006EA 183.08 USD
101912-006 101912-008
2,4-Dichlorobenzhydrazide
2,4-Dichlorobenzhydrazide

Hazard code(s): Irritant

Formula: C₇H₆Cl₂N₂O
MW: 205.04 g/mol
Melting Pt: 168…170 °C
MDL Number: MFCD00025113
CAS Number: 5814-06-2

Order Now

Specification Test Results

Melting point 168-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR