2,4-Dichloroanisole ≥99%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloro-1-methoxybenzene

056997-10G 056997-50G
101912-002EA 95.38 USD
101912-002 101912-004
2,4-Dichloroanisole ≥99%
2,4-Dichloroanisole

Hazard code(s): Irritant

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 125 °C (10mm)
Melting Pt: 28…29 °C
Density: 1.26 g/cm³ (20 °C)
MDL Number: MFCD00044772
CAS Number: 553-82-2
EINECS: 209-051-5

Order Now

Specification Test Results

Boiling point 125°C/10mm
Melting point 28-29°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR