3,4-Dibromotoluene ≥99%

Supplier: Matrix Scientific

Synonyms: 1,2-Dibromo-4-methylbenzene

056995-5G 056995-25G 056995-100G
101911-994EA 80 USD
101911-994 101911-996 101911-998
3,4-Dibromotoluene ≥99%
3,4-Dibromotoluene

Order Now

Specification Test Results

Boiling point 112°C/20mm
Density 1.83

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR