Cyclohexyltriphenylphosphonium bromide for synthesis

Supplier: Matrix Scientific

056990-10G 056990-50G 056990-250G
101911-980EA 192.31 USD
101911-980 101911-982 101911-984
Cyclohexyltriphenylphosphonium bromide for synthesis
Cyclohexyltriphenylphosphonium bromide

Hazard code(s): Irritant, Hygroscopic

Formula: C₂₄H₂₆BrP
MW: 425.35
Melting Pt: 270…272 °C
MDL Number: MFCD00031667
CAS Number: 7333-51-9
EINECS: 230-834-2

Order Now

Specification Test Results

Melting point 270-272°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR