3-Chlorostyrene ≥99% stabilized

Supplier: Matrix Scientific

056985-5G 056985-25G
101911-964EA 67.69 USD
101911-964 101911-966
3-Chlorostyrene ≥99% stabilized
3-Chlorostyrene

Inhibited


Hazard code(s): Irritant, Refrigerate

Formula: C₈H₇Cl
Boiling Pt: 62…63 °C (6mm)
Melting Pt: –26 °C
Density: 1.09 g/cm³
MDL Number: MFCD00000598
CAS Number: 2039-85-2
EINECS: 218-024-7

Order Now

Specification Test Results

Boiling point 62-63°C/6mm
Density 1.09


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR